- The receptor subunit Tom20 is dynamically associated with the TOM complex in mitochondria of human cells.
The receptor subunit Tom20 is dynamically associated with the TOM complex in mitochondria of human cells.
The outer membrane translocase (TOM) is the import channel for nuclear-encoded mitochondrial proteins. The general import pore contains Tom40, Tom22, Tom5, Tom6, and Tom7. Precursor proteins are bound by the (peripheral) receptor proteins Tom20, Tom22, and Tom70 before being imported by the TOM complex. Here we investigated the association of the receptor Tom20 with the TOM complex. Tom20 was found in the TOM complex, but not in a smaller subcomplex. In addition, a subcomplex was found without Tom40 and Tom7 but with Tom20. Using single particle tracking of labeled Tom20 in overexpressing human cells, we show that Tom20 has, on average, higher lateral mobility in the membrane than Tom7/TOM. After ligation of Tom20 with the TOM complex by post-tranlational protein trans-splicing using the traceless, ultrafast cleaved Gp41-1 integrin system, a significant decrease in the mean diffusion coefficient of Tom20 was observed in the resulting Tom20-Tom7 fusion protein. Exposure of Tom20 to high substrate loading also resulted in reduced mobility. Taken together, our data show that the receptor subunit Tom20 interacts dynamically with the TOM core complex. We suggest that the TOM complex containing Tom20 is the active import pore and that Tom20 is associated when substrate is available.