MilliporeSigma
Monoclonal anti-A activity against the FORS1 (Forssman) antigen.
Transfusion (2014-07-22)
Katie Barr, Elena Korchagina, Inna Popova, Nicolai Bovin, Stephen Henry
ABSTRACT
The FORS blood group system (originally recognized as the Apae phenotype) was discovered by sporadic activity against polyclonal anti-A reagents and activity against the lectin Helix pomatia. The extent of monoclonal anti-A reagent activity against the FORS1 antigen is serologically and immunochemically incomplete. In the absence of natural FORS1-positive red blood cells (RBCs), kodecytes were created with synthetic disaccharide and pentasaccharide Forssman function-spacer-lipid (FSL) constructs, Fsdi -kodecytes, and FORS1-kodecytes, respectively. FSL constructs were also applied to solid surfaces and used in solid-phase enzyme immunoassays. A range of characterized monoclonal anti-A and anti-B reagents were then serologically and immunochemically characterized against these Forssman antigens. Polyclonal human anti-A, anti-B, the lectin H. pomatia serologic reagents; and canine RBCs were used as serologic controls. None of 19 different monoclonal anti-A reagents were able to detect the pentasaccharide Forssman on FORS1-kodecytes, while three reagents were able to detect disaccharide Forssman on Fsdi -kodecytes. Most anti-A reagents were immunochemically reactive with both the di- and the pentasaccharide Forssman antigens in the solid-phase assays. Historic polyclonal human anti-A and the lectin H. pomatia reacted strongly with the FORS1-kodecytes, correlating with the discovery of the Apae phenotype and supporting the use of FORS1-kodecytes as FORS1 surrogates. Monoclonal anti-A reagents, despite showing reactivity against the FORS1 antigen in solid-phase assays are unlikely to cause the agglutination of FORS1 antigen-positive RBCs.
Sigma-Aldrich
Lectin from Helix pomatia, lyophilized powder, salt, free
Pricing and availability is not currently available.