MilliporeSigma
  • Homozygous nonsense mutation in SCHIP1/IQCJ-SCHIP1 causes a neurodevelopmental brain malformation syndrome.
Homozygous nonsense mutation in SCHIP1/IQCJ-SCHIP1 causes a neurodevelopmental brain malformation syndrome.
Clinical genetics (2017-08-09)
M F Elsaid, N Chalhoub, T Ben-Omran, H Kamel, M Al Mureikhi, K Ibrahim, M Elizabeth Ross, A K Abdel Aleem
ABSTRACT
We report a consanguineous Arab family with 3 affected siblings who display a disorder of global developmental delay, learning difficulties, facial dysmorphism, hearing impairments, and cataract. The clinical phenotype was associated with characteristic brain magnetic resonance imaging (MRI) features of axonal guidance defects involving anterior commissure agenesis as well as scattered areas of polymicrogyria-cobblestone complex. Whole genome sequencing revealed a novel nonsense mutation (159609921C>T) that segregated in the family consistent in an autosomal recessive pattern. This mutation located in the C-terminal region shared by the Schwanomin-Interacting Protein1 (SCHIP1) isoforms including the IQCJ-SCHIP1. The in vitro expression of SCHIP1 and IQCJ-SCHIP1 truncated mutant isoforms (NM_001197109.1; p.R209* and NM_001197114.1; p.R501*, respectively) were markedly reduced as compared to their full-length versions suggesting protein stability/folding impairment. The pathogenic nature of this mutation is supported by a previously reported mouse knockout of Schip1 isoforms, which phenocopied the human axon guidance abnormality. This is the first report of a SCHIP1/IQCJ-SCHIP1 point mutation in humans associated with a neurological-developmental phenotype.
Sigma-Aldrich
Anti-SCHIP1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution