MilliporeSigma
Folium Sennae and emodin reverse airway smooth muscle contraction.
Cell biology international (2020-05-22)
Jun-Ying Qiu, Li-Qun Ma, Bei-Bei Liu, Wen-Jing Zhang, Meng-Su Liu, Ge-Ge Wang, Xiao-Xue Zhao, Xi Luo, Qian Wang, Hao Xu, Dun-An Zang, Jinhua Shen, Yong-Bo Peng, Ping Zhao, Lu Xue, Meng-Fei Yu, Weiwei Chen, Jiapei Dai, Qing-Hua Liu
ABSTRACT
The objective of this project was to find a bronchodilatory compound from herbs and clarify the mechanism. We found that the ethanol extract of Folium Sennae (EEFS) can relax airway smooth muscle (ASM). EEFS inhibited ASM contraction, induced by acetylcholine, in mouse tracheal rings and lung slices. High-performance liquid chromatography assay showed that EEFS contained emodin. Emodin had a similar reversal action. Acetylcholine-evoked contraction was also partially reduced by nifedipine (a selective inhibitor of L-type voltage-dependent Ca2+ channels, LVDCCs), YM-58483 (a selective inhibitor of store-operated Ca2+ entry, SOCE), as well as Y-27632 (an inhibitor of Rho-associated protein kinase). In addition, LVDCC- and SOCE-mediated currents and cytosolic Ca2+ elevations were inhibited by emodin. Emodin reversed acetylcholine-caused increases in phosphorylation of myosin phosphatase target subunit 1. Furthermore, emodin, in vivo, inhibited acetylcholine-induced respiratory system resistance in mice. These results indicate that EEFS-induced relaxation results from emodin inhibiting LVDCC, SOCE, and Ca2+ sensitization. These findings suggest that Folium Sennae and emodin may be new sources of bronchodilators.