MilliporeSigma
  • Discovery and Characterization of the Potent and Highly Selective 1,7-Naphthyridine-Based Inhibitors BAY-091 and BAY-297 of the Kinase PIP4K2A.
Discovery and Characterization of the Potent and Highly Selective 1,7-Naphthyridine-Based Inhibitors BAY-091 and BAY-297 of the Kinase PIP4K2A.
Journal of medicinal chemistry (2021-10-27)
Lars Wortmann, Nico Bräuer, Simon J Holton, Horst Irlbacher, Jörg Weiske, Christian Lechner, Robin Meier, Jakob Karén, Catrine Berthold Siöberg, Vera Pütter, Clara D Christ, Antonius Ter Laak, Philip Lienau, Ralf Lesche, Barbara Nicke, Shing-Hu Cheung, Marcus Bauser, Andrea Haegebarth, Franz von Nussbaum, Dominik Mumberg, Clara Lemos
ABSTRACT
PIP4K2A is an insufficiently studied type II lipid kinase that catalyzes the conversion of phosphatidylinositol-5-phosphate (PI5P) into phosphatidylinositol 4,5-bisphosphate (PI4,5P2). The involvement of PIP4K2A/B in cancer has been suggested, particularly in the context of p53 mutant/null tumors. PIP4K2A/B depletion has been shown to induce tumor growth inhibition, possibly due to hyperactivation of AKT and reactive oxygen species-mediated apoptosis. Herein, we report the identification of the novel potent and highly selective inhibitors BAY-091 and BAY-297 of the kinase PIP4K2A by high-throughput screening and subsequent structure-based optimization. Cellular target engagement of BAY-091 and BAY-297 was demonstrated using cellular thermal shift assay technology. However, inhibition of PIP4K2A with BAY-091 or BAY-297 did not translate into the hypothesized mode of action and antiproliferative activity in p53-deficient tumor cells. Therefore, BAY-091 and BAY-297 serve as valuable chemical probes to study PIP4K2A signaling and its involvement in pathophysiological conditions such as cancer.
Sigma-Aldrich
BAY-0361, ≥95% (HPLC)
Sigma-Aldrich
BAY-091, ≥98% (HPLC)
Sigma-Aldrich
Fetal Bovine Serum, USA origin, sterile-filtered, suitable for cell culture, suitable for hybridoma