MilliporeSigma
  • Ghrelin gene products rescue cultured adult rat hippocampal neural stem cells from high glucose insult.
Ghrelin gene products rescue cultured adult rat hippocampal neural stem cells from high glucose insult.
Journal of molecular endocrinology (2016-08-18)
Sehee Kim, Chanyang Kim, Seungjoon Park
ABSTRACT
Adult hippocampal neurogenesis is decreased in type 2 diabetes, and this impairment appears to be important in cognitive dysfunction. Previous studies suggest that ghrelin gene products (acylated ghrelin (AG), unacylated ghrelin (UAG) and obestatin (OB)) promote neurogenesis. Therefore, we hypothesize that ghrelin gene products may reduce the harmful effects of high glucose (HG) on hippocampal neural stem cells (NSCs). The aim of this study was to investigate the role of these peptides on the survival of cultured hippocampal NSCs exposed to HG insult. Treatment of hippocampal NSCs with AG, UAG or OB inhibited HG-induced cell death and apoptosis. Exposure of cells to the growth hormone secretagogue receptor 1a antagonist abolished the protective effects of AG against HG toxicity, whereas those of UAG or OB were preserved. All three peptides attenuated HG-induced decrease in BrdU-labeled and phosphohistone-H3-labeled cells. We also investigated the effects of ghrelin gene products on the regulation of apoptosis at the mitochondrial level. AG, UAG or OB rescued hippocampal NSCs from HG insult by inhibiting intracellular and mitochondrial reactive oxygen species generation and stabilizing mitochondrial transmembrane potential. In addition, cells treated with ghrelin gene products showed an increased Bcl-2 and decreased Bax levels, thereby increasing the Bcl-2/Bax ratio, inhibiting cytochrome c release and preventing caspase-3 activation. Finally, AG-, UAG- or OB-mediated protection was dependent on the activities of adenosine monophosphate-activated protein kinase/uncoupling protein 2 pathway. Our data indicate that ghrelin gene products may act as survival factors that preserve mitochondrial function and inhibit oxidative stress-induced apoptosis.
Sigma-Aldrich
Adult Rat Hippocampal Neural Stem Cells, Adult Rat Hippocampal Neural Stem Cells are self-renewing cells with the capacity to differentiate into neurons, astrocytes & oligodendrocytes.
Pricing and availability is not currently available.