MilliporeSigma
  • Clinical Features and Multiplatform Molecular Analysis Assist in Understanding Patient Response to Anti-PD-1/PD-L1 in Renal Cell Carcinoma.
Clinical Features and Multiplatform Molecular Analysis Assist in Understanding Patient Response to Anti-PD-1/PD-L1 in Renal Cell Carcinoma.
Cancers (2021-04-04)
Eileen Shiuan, Anupama Reddy, Stephanie O Dudzinski, Aaron R Lim, Ayaka Sugiura, Rachel Hongo, Kirsten Young, Xian-De Liu, Christof C Smith, Jamye O'Neal, Kimberly B Dahlman, Renee McAlister, Beiru Chen, Kristen Ruma, Nathan Roscoe, Jehovana Bender, Joolz Ward, Ju Young Kim, Christine Vaupel, Jennifer Bordeaux, Shridar Ganesan, Tina M Mayer, Gregory M Riedlinger, Benjamin G Vincent, Nancy B Davis, Scott M Haake, Jeffrey C Rathmell, Eric Jonasch, Brian I Rini, W Kimryn Rathmell, Kathryn E Beckermann
ABSTRACT
Predicting response to ICI therapy among patients with renal cell carcinoma (RCC) has been uniquely challenging. We analyzed patient characteristics and clinical correlates from a retrospective single-site cohort of advanced RCC patients receiving anti-PD-1/PD-L1 monotherapy (N = 97), as well as molecular parameters in a subset of patients, including multiplexed immunofluorescence (mIF), whole exome sequencing (WES), T cell receptor (TCR) sequencing, and RNA sequencing (RNA-seq). Clinical factors such as the development of immune-related adverse events (odds ratio (OR) = 2.50, 95% confidence interval (CI) = 1.05-5.91) and immunological prognostic parameters, including a higher percentage of circulating lymphocytes (23.4% vs. 17.4%, p = 0.0015) and a lower percentage of circulating neutrophils (61.8% vs. 68.5%, p = 0.0045), correlated with response. Previously identified gene expression signatures representing pathways of angiogenesis, myeloid inflammation, T effector presence, and clear cell signatures also correlated with response. High PD-L1 expression (>10% cells) as well as low TCR diversity (≤644 clonotypes) were associated with improved progression-free survival (PFS). We corroborate previously published findings and provide preliminary evidence of T cell clonality impacting the outcome of RCC patients. To further biomarker development in RCC, future studies will benefit from integrated analysis of multiple molecular platforms and prospective validation.
Sigma-Aldrich
Anti-CD25 antibody, Rabbit monoclonal, recombinant, expressed in proprietary host, clone SP176, affinity isolated antibody