MilliporeSigma
  • Age-dependent effects of the onset of a conductive hearing loss on the volume of the cochlear nucleus subdivisions and the expression of c-fos in the mongolian gerbil (Meriones unguiculatus).
Age-dependent effects of the onset of a conductive hearing loss on the volume of the cochlear nucleus subdivisions and the expression of c-fos in the mongolian gerbil (Meriones unguiculatus).
Audiology & neuro-otology (1997-05-01)
O Gleich, J Strutz
ABSTRACT
A monaural conductive hearing loss was induced by interrupting the chain of the middle ear ossicles on the right side in gerbils of four different age groups (P12-14, P20-21, P42 and P84). The volumes of the cochlear nucleus subdivisions and the number of cells that expressed immunoreactivity for c-fos after noise stimulation were determined on the left and right side in the deprived animals, and in undeprived control animals when they reached the age of 6 months. The anteroventral cochlear nucleus on the deprived side was reduced in volume when the deprivation started before the age of 3 months. The other cochlear nucleus subdivisions showed no systematic age-dependent reductions. The expression of c-fos in the dorsal cochlear nucleus appeared more resistant to a hearing loss, with deprivation being more effective in younger animals. c-fos expression was also dramatically reduced in the ventral cochlear nucleus, regardless of age at the onset of hearing loss.