MilliporeSigma
DIPA-CRISPR is a simple and accessible method for insect gene editing.
Cell reports methods (2022-06-01)
Yu Shirai, Maria-Dolors Piulachs, Xavier Belles, Takaaki Daimon
ABSTRACT
Current approaches for insect gene editing require microinjection of materials into early embryos. This severely limits the application of gene editing to a great number of insect species, especially to those whose reproduction systems preclude access to early embryos for injection. To overcome these limitations, we report a simple and accessible method for insect gene editing, termed "direct parental" CRISPR (DIPA-CRISPR). We show that injection of Cas9 ribonucleoproteins (RNPs) into the haemocoel of adult females efficiently introduces heritable mutations in developing oocytes. Importantly, commercially available standard Cas9 protein can be directly used for DIPA-CRISPR, which makes this approach highly practical and feasible. DIPA-CRISPR enables highly efficient gene editing in the cockroaches, on which conventional approaches cannot be applied, and in the model beetle Tribolium castaneum. Due to its simplicity and accessibility, DIPA-CRISPR will greatly extend the application of gene editing technology to a wide variety of insects.
Sigma-Aldrich
Cas9 Protein, from Streptococcus pyogenes, recombinant, expressed in E. coli, 1X NLS
Pricing and availability is not currently available.