MilliporeSigma
  • Phosphate-mediated coanchoring of RBD immunogens and molecular adjuvants to alum potentiates humoral immunity against SARS-CoV-2.
Phosphate-mediated coanchoring of RBD immunogens and molecular adjuvants to alum potentiates humoral immunity against SARS-CoV-2.
Science advances (2021-12-09)
Kristen A Rodrigues, Sergio A Rodriguez-Aponte, Neil C Dalvie, Jeong Hyun Lee, Wuhbet Abraham, Diane G Carnathan, Luis E Jimenez, Julia T Ngo, Jason Y H Chang, Zeli Zhang, Jingyou Yu, Aiquan Chang, Catherine Nakao, Benjamin Goodwin, Christopher A Naranjo, Libin Zhang, Murillo Silva, Dan H Barouch, Guido Silvestri, Shane Crotty, J Christopher Love, Darrell J Irvine
ABSTRACT
There is a need for additional rapidly scalable, low-cost vaccines against severe acute respiratory syndrome coronavirus 2 (SARS-CoV-2) to achieve global vaccination. Aluminum hydroxide (alum) adjuvant is the most widely available vaccine adjuvant but elicits modest humoral responses. We hypothesized that phosphate-mediated coanchoring of the receptor binding domain (RBD) of SARS-CoV-2 together with molecular adjuvants on alum particles could potentiate humoral immunity by promoting extended vaccine kinetics and codelivery of vaccine components to lymph nodes. Modification of RBD immunogens with phosphoserine (pSer) peptides enabled efficient alum binding and slowed antigen clearance, leading to notable increases in germinal center responses and neutralizing antibody titers in mice. Adding phosphate-containing CpG or saponin adjuvants to pSer-RBD:alum immunizations synergistically enhanced vaccine immunogenicity in mice and rhesus macaques, inducing neutralizing responses against SARS-CoV-2 variants. Thus, phosphate-mediated coanchoring of RBD and molecular adjuvants to alum is an effective strategy to enhance the efficacy of SARS-CoV-2 subunit vaccines.
Sigma-Aldrich
Cytochrome c from Saccharomyces cerevisiae, ≥85% based on Mol. Wt. 12,588 basis
Sigma-Aldrich
Kaiser test kit