MilliporeSigma
  • Human and mouse single-nucleus transcriptomics reveal TREM2-dependent and TREM2-independent cellular responses in Alzheimer's disease.
Human and mouse single-nucleus transcriptomics reveal TREM2-dependent and TREM2-independent cellular responses in Alzheimer's disease.
Nature medicine (2020-01-15)
Yingyue Zhou, Wilbur M Song, Prabhakar S Andhey, Amanda Swain, Tyler Levy, Kelly R Miller, Pietro L Poliani, Manuela Cominelli, Shikha Grover, Susan Gilfillan, Marina Cella, Tyler K Ulland, Konstantin Zaitsev, Akinori Miyashita, Takeshi Ikeuchi, Makoto Sainouchi, Akiyoshi Kakita, David A Bennett, Julie A Schneider, Michael R Nichols, Sean A Beausoleil, Jason D Ulrich, David M Holtzman, Maxim N Artyomov, Marco Colonna
ABSTRACT
Glia have been implicated in Alzheimer's disease (AD) pathogenesis. Variants of the microglia receptor triggering receptor expressed on myeloid cells 2 (TREM2) increase AD risk, and activation of disease-associated microglia (DAM) is dependent on TREM2 in mouse models of AD. We surveyed gene-expression changes associated with AD pathology and TREM2 in 5XFAD mice and in human AD by single-nucleus RNA sequencing. We confirmed the presence of Trem2-dependent DAM and identified a previously undiscovered Serpina3n+C4b+ reactive oligodendrocyte population in mice. Interestingly, remarkably different glial phenotypes were evident in human AD. Microglia signature was reminiscent of IRF8-driven reactive microglia in peripheral-nerve injury. Oligodendrocyte signatures suggested impaired axonal myelination and metabolic adaptation to neuronal degeneration. Astrocyte profiles indicated weakened metabolic coordination with neurons. Notably, the reactive phenotype of microglia was less evident in TREM2-R47H and TREM2-R62H carriers than in non-carriers, demonstrating a TREM2 requirement in both mouse and human AD, despite the marked species-specific differences.
Sigma-Aldrich
HOG Human Oligodendroglioma Cell Line, HOG human oligodendroglioma cell line is a useful model to study the role of oligodendrocytes in neurodegenerative diseases.