MilliporeSigma
Unconventional secretion of FABP4 by endosomes and secretory lysosomes.
The Journal of cell biology (2017-12-08)
Julien Villeneuve, Laia Bassaganyas, Sebastien Lepreux, Marioara Chiritoiu, Pierre Costet, Jean Ripoche, Vivek Malhotra, Randy Schekman
ABSTRACT
An appreciation of the functional properties of the cytoplasmic fatty acid binding protein 4 (FABP4) has advanced with the recent demonstration that an extracellular form secreted by adipocytes regulates a wide range of physiological functions. Little, however, is known about the mechanisms that mediate the unconventional secretion of FABP4. Here, we demonstrate that FABP4 secretion is mediated by a membrane-bounded compartment, independent of the conventional endoplasmic reticulum-Golgi secretory pathway. We show that FABP4 secretion is also independent of GRASP proteins, autophagy, and multivesicular bodies but involves enclosure within endosomes and secretory lysosomes. We highlight the physiological significance of this pathway with the demonstration that an increase in plasma levels of FABP4 is inhibited by chloroquine treatment of mice. These findings chart the pathway of FABP4 secretion and provide a potential therapeutic means to control metabolic disorders associated with its dysregulated secretion.
Sigma-Aldrich
Golgi Isolation Kit, sufficient for 50 g (tissue)
Pricing and availability is not currently available.