MilliporeSigma
  • Targeting the PI3K pathway in the brain--efficacy of a PI3K inhibitor optimized to cross the blood-brain barrier.
Targeting the PI3K pathway in the brain--efficacy of a PI3K inhibitor optimized to cross the blood-brain barrier.
Clinical cancer research : an official journal of the American Association for Cancer Research (2012-09-21)
Laurent Salphati, Timothy P Heffron, Bruno Alicke, Merry Nishimura, Kai Barck, Richard A Carano, Jonathan Cheong, Kyle A Edgar, Joan Greve, Samir Kharbanda, Hartmut Koeppen, Shari Lau, Leslie B Lee, Jodie Pang, Emile G Plise, Jenny L Pokorny, Hani Bou Reslan, Jann N Sarkaria, Jeffrey J Wallin, Xiaolin Zhang, Stephen E Gould, Alan G Olivero, Heidi S Phillips
ABSTRACT
Glioblastoma (GBM), the most common primary brain tumor in adults, presents a high frequency of alteration in the PI3K pathway. Our objectives were to identify a dual PI3K/mTOR inhibitor optimized to cross the blood-brain barrier (BBB) and characterize its brain penetration, pathway modulation in the brain and efficacy in orthotopic xenograft models of GBM. Physicochemical properties of PI3K inhibitors were optimized using in silico tools, leading to the identification of GNE-317. This compound was tested in cells overexpressing P-glycoprotein (P-gp) or breast cancer resistance protein (BCRP). Following administration to mice, GNE-317 plasma and brain concentrations were determined, and phosphorylated biomarkers (pAkt, p4EBP1, and pS6) were measured to assess PI3K pathway suppression in the brain. GNE-317 efficacy was evaluated in the U87, GS2, and GBM10 orthotopic models of GBM. GNE-317 was identified as having physicochemical properties predictive of low efflux by P-gp and BCRP. Studies in transfected MDCK cells showed that GNE-317 was not a substrate of either transporter. GNE-317 markedly inhibited the PI3K pathway in mouse brain, causing 40% to 90% suppression of the pAkt and pS6 signals up to 6-hour postdose. GNE-317 was efficacious in the U87, GS2, and GBM10 orthotopic models, achieving tumor growth inhibition of 90% and 50%, and survival benefit, respectively. These results indicated that specific optimization of PI3K inhibitors to cross the BBB led to potent suppression of the PI3K pathway in healthy brain. The efficacy of GNE-317 in 3 intracranial models of GBM suggested that this compound could be effective in the treatment of GBM.
Sigma-Aldrich
GNE-317, ≥98% (HPLC)