MilliporeSigma
  • Modified aptamers enable quantitative sub-10-nm cellular DNA-PAINT imaging.
Modified aptamers enable quantitative sub-10-nm cellular DNA-PAINT imaging.
Nature methods (2018-08-22)
Sebastian Strauss, Philipp C Nickels, Maximilian T Strauss, Vilma Jimenez Sabinina, Jan Ellenberg, Jeffrey D Carter, Shashi Gupta, Nebojsa Janjic, Ralf Jungmann
ABSTRACT
Although current implementations of super-resolution microscopy are technically approaching true molecular-scale resolution, this has not translated to imaging of biological specimens, because of the large size of conventional affinity reagents. Here we introduce slow off-rate modified aptamers (SOMAmers) as small and specific labeling reagents for use with DNA points accumulation in nanoscale topography (DNA-PAINT). To demonstrate the achievable resolution, specificity, and multiplexing capability of SOMAmers, we labeled and imaged both transmembrane and intracellular targets in fixed and live cells.
Product Description
Millipore
Green Fluorescent Protein