MilliporeSigma
  • MRTF-A regulates myoblast commitment to differentiation by targeting PAX7 during muscle regeneration.
MRTF-A regulates myoblast commitment to differentiation by targeting PAX7 during muscle regeneration.
Journal of cellular and molecular medicine (2021-08-05)
Ruhui Song, Shengnan Zhao, Yue Xu, Jian Hu, Shuangao Ke, Fan Li, Gaohui Tian, Xiao Zheng, Jiajun Li, Lixing Gu, Yao Xu
ABSTRACT
Myocardin-related transcription factor-A/serum response factor (MRTF-A/SRF), a well-known transcriptional programme, has been proposed to play crucial roles in skeletal muscle development and function. However, whether MRTF-A participates in muscle regeneration and the molecular mechanisms are not completely understood. Here, we show that MRTF-A levels are highly correlated with myogenic genes using a RNA-seq assay, which reveal that MRTF-A knockdown in C2C12 cells significantly reduces PAX7 expression. Subsequent in vitro and in vivo data show that MRTF-A and PAX7 present identical expression patterns during myoblast differentiation and CTX-induced muscle injury and repair. Remarkably, MRTF-A overexpression promotes myoblast proliferation, while inhibiting cell differentiation and the expression of MyoD and MyoG. MRTF-A loss of function produces the opposite effect. Moreover, mice with lentivirus (MRTF-A) injection possesses more PAX7+ satellite cells, but less differentiating MyoD+ and MyoG+ cells, leading subsequently to diminished muscle regeneration. Our mechanistic results reveal that MRTF-A contributes to PAX7-mediated myoblast self-renewal, proliferation, and differentiation by binding to its distal CArG box. Overall, we propose that MRTF-A functions as a novel PAX7 regulator upon myoblast commitment to differentiation, which could provide pathways for dictating muscle stem cell fate and open new avenues to explore stem cell-based therapy for muscle degenerative diseases.
Sigma-Aldrich
Monoclonal Anti-Myogenin antibody produced in mouse, clone F12B, purified immunoglobulin, buffered aqueous solution