MilliporeSigma
  • Modafinil Reduces Neuronal Pyroptosis and Cognitive Decline After Sleep Deprivation.
Modafinil Reduces Neuronal Pyroptosis and Cognitive Decline After Sleep Deprivation.
Frontiers in neuroscience (2022-03-22)
Xiangyang Xiong, Yan Zuo, Lu Cheng, Zhenyu Yin, Tianpeng Hu, Mengtian Guo, Zhaoli Han, Xintong Ge, Wenzhu Li, Yan Wang, Dong Wang, Conglin Wang, Lan Zhang, Yaodan Zhang, Qiang Liu, Fanglian Chen, Ping Lei
ABSTRACT
Sleep deprivation (SD) induces systemic inflammation that promotes neuronal pyroptosis. The purpose of this study was to investigate the effect of an antioxidant modafinil on neuronal pyroptosis and cognitive decline following SD. Using a mouse model of SD, we found that modafinil improved learning and memory, reduced proinflammatory factor (IL-1β, TNF-α, and IL-6) production, and increased the expression of anti-inflammatory factors (IL-10). Modafinil treatment attenuated inflammasome activity and reduced neuronal pyroptosis involving the NLRP3/NLRP1/NLRC4-caspase-1-IL-1β pathway. In addition, modafinil induced an upregulation of brain-derived neurotrophic factor (BDNF) and synaptic activity. These results suggest that modafinil reduces neuronal pyroptosis and cognitive decline following SD. These effects should be further investigated in future studies to benefit patients with sleep disorders.
Sigma-Aldrich
Anti-Ipaf (NLRC4) Antibody, from rabbit, purified by affinity chromatography