MilliporeSigma
A Review on Medicinal Properties of Orientin.
Advances in pharmacological sciences (2016-06-15)
Kit Ying Lam, Anna Pick Kiong Ling, Rhun Yian Koh, Ying Pei Wong, Yee How Say
ABSTRACT
Medicinal plants continue to play an important role in modern medications and healthcare as consumers generally believe that most of them cause fewer or milder adverse effects than the conventional modern medicines. In order to use the plants as a source of medicinal agents, the bioactive compounds are usually extracted from plants. Therefore, the extraction of bioactive compounds from medicinal plants is a crucial step in producing plant-derived drugs. One of the bioactive compounds isolable from medicinal plants, orientin, is often used in various bioactivity studies due to its extensive beneficial properties. The extraction of orientin in different medicinal plants and its medicinal properties, which include antioxidant, antiaging, antiviral, antibacterial, anti-inflammation, vasodilatation and cardioprotective, radiation protective, neuroprotective, antidepressant-like, antiadipogenesis, and antinociceptive effects, are discussed in detail in this review.
Sigma-Aldrich
Orientin, ≥97% (HPLC)