MilliporeSigma
  • Fascin enhances the vulnerability of breast cancer to erastin-induced ferroptosis.
Fascin enhances the vulnerability of breast cancer to erastin-induced ferroptosis.
Cell death & disease (2022-02-16)
Cong Chen, Bojian Xie, Zhaoqing Li, Lini Chen, Yongxia Chen, Jichun Zhou, Siwei Ju, Yulu Zhou, Xun Zhang, Wenying Zhuo, Jingjing Yang, Misha Mao, Ling Xu, Linbo Wang
ABSTRACT
Ferroptosis, which is characterized by intracellular iron accumulation and lipid peroxidation, is a newly described form of regulated cell death that may play a key role in tumour suppression. In the present study, we investigated the expression profiles and biological effects of fascin actin-bundling protein 1 (Fascin, gene name FSCN1) in breast cancer. In addition, bioinformatics analysis of the TCGA cancer database and gain- and loss-of-function studies showed that Fascin enhances sensitivity to erastin-induced ferroptosis. Mechanistically, Fascin directly interacts with cysteine/glutamate transporter (xCT, gene name SLC7A11) and decreases its stability via the ubiquitin-mediated proteasome degradation pathway. Furthermore, we observed that Fascin is substantially upregulated in tamoxifen-resistant breast cancer cell lines, and drug-resistant cells were also more vulnerable to erastin-induced ferroptosis. Taken together, our findings reveal a previously unidentified role of Fascin in ferroptosis by regulating xCT. Thus, ferroptosis activation in breast cancer with high Fascin level may serve as a potential treatment.