MilliporeSigma
  • Loss of CREBRF Reduces Anxiety-like Behaviors and Circulating Glucocorticoids in Male and Female Mice.
Loss of CREBRF Reduces Anxiety-like Behaviors and Circulating Glucocorticoids in Male and Female Mice.
Endocrinology (2020-09-10)
Krystle A Frahm, Akeem A Williams, Ashlee N Wood, Michael C Ewing, Polly E Mattila, Byron W Chuan, Lanping Guo, Faraaz A Shah, Christopher P O'Donnell, Ray Lu, Donald B DeFranco
ABSTRACT
Glucocorticoid signaling controls many key biological functions ranging from stress responses to affective states. The putative transcriptional coregulator CREB3 regulatory factor (CREBRF) reduces glucocorticoid receptor levels in vitro, suggesting that CREBRF may impact behavioral and physiological outputs. In the present study, we examined adult male and female mice with global loss of CREBRF (CrebrfKO) for anxiety-like behaviors and circulating glucocorticoids in response to various acute stress conditions. Results demonstrate that both male and female CrebrfKO mice have preserved locomotor activity but reduced anxiety-like behaviors during the light-dark box and elevated plus maze. These behavioral phenotypes were associated with lower plasma corticosterone after restraint stress. Further studies using unhandled female mice also demonstrated a loss of the diurnal circulating corticosterone rhythm in CrebrfKO mice. These results suggest that CREBRF impacts anxiety-like behavior and circulating glucocorticoids in response to acute stressors and serves as a basis for future mechanistic studies to define the impact of CREBRF in glucocorticoid-associated behavioral and physiological responses.
Sigma-Aldrich
Corticotropin Releasing Factor human, rat, ≥95% (HPLC), powder