MilliporeSigma
  • Gold(I) and Silver(I) Complexes with 2-Anilinopyridine-Based Heterocycles as Multitarget Drugs against Colon Cancer.
Gold(I) and Silver(I) Complexes with 2-Anilinopyridine-Based Heterocycles as Multitarget Drugs against Colon Cancer.
Inorganic chemistry (2020-11-19)
Inés Mármol, Sara Montanel-Perez, José Carlos Royo, M Concepción Gimeno, M Dolores Villacampa, M Jesús Rodríguez-Yoldi, Elena Cerrada
ABSTRACT
A series of gold(I) and silver(I) derivatives with N- or S-donor ligands derived from 2-anilinopyridine has been synthesized and characterized. The mononuclear structure of [Au(L1)(PPh3)](TfO) (1a) and [Au(L2)(PPh3)](TfO) (1b) was confirmed by X-ray diffraction studies, as well as the dinuclear structure in the case of [Ag(TfO)(L1)]2 (4a). Most of the complexes are cytotoxic against a model of colorectal adenocarcinoma (Caco-2 cell line) and breast adenocarcinoma cancer cell lines (MCF-7). [Au(L1)(PPh3)](TfO) (1a) was able to induce caspases 8 and 3 activation, loss of mitochondrial membrane potential, and reactive oxygen species (ROS)-dependent cell death on Caco-2 cells upon 24 h incubation. In addition, the gold complex 1a produced a significant inhibition of the redox enzyme thioredoxin reductase as well as 20S proteasome. However, the silver(I) analogue, [Ag(TfO)(L1)(PPh3)] (2a), induced cell death independent of inhibition of thioredoxin reductase and 20S proteasome, triggered ROS-independent apoptosis mediated by caspase 8 and 3 activation, and loss of mitochondrial membrane potential, which points to a different mechanism of action for both derivatives, dependent on the metal center.
Sigma-Aldrich
Proteasome 20S Activity Assay Kit, sufficient for 100 fluorometric tests