MilliporeSigma
  • Silica-based magnetic hybrid nanocomposite for the extraction and preconcentration of some organophosphorus pesticides before gas chromatography.
Silica-based magnetic hybrid nanocomposite for the extraction and preconcentration of some organophosphorus pesticides before gas chromatography.
Journal of separation science (2018-06-08)
Somayeh Koupaei Malek, Hamid Rashidi Nodeh, Behrouz Akbari-Adergani
ABSTRACT
The precise control of pesticide residues in foodstuffs depends significantly on the clean extraction of analytes using specifically designed separation methods. In this study, a one-pot sol-gel process was used for the preparation of a magnetic hybrid silica gel tetraethylortho silicate-cyanopropyltriethoxy silane nanocomposite. The prepared material was characterized using energy dispersive X-ray spectroscopy, Fourier transform infrared spectroscopy, single-point specific surface area, and scanning electron microcopy. The synthesized magnetic hybrid material was used as a solid phase extraction sorbent for the extraction and preconcentration of some organophosphorus pesticides before gas chromatography with a microelectron capture detector. The performance of the proposed magnetic solid-phase extraction technique was validated by linearity (0.05-2 ng/mL), correlation coefficients (r2  = 0.9993-0.9997), limit of detection (0.02-0.06 ng/mL, S/N = 3, n = 3), and intraday (RSD = 1.5-8.7%, n = 3) and interday precision (RSD = 5.5-9.3%, n = 12), while the recovery in real samples and equilibrium adsorption capacity was 72.02-103.84% and 8-20 mg/g, respectively. The magnetic solid-phase extraction based on the hybrid nanocomposite revealed a high enrichment factor, an appropriate dynamic range, and great absorptive ability toward the selected organophosphorus pesticides spiked in real water samples.
Product Description
Supelco
Phosphamidon, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Pricing and availability is not currently available.