MilliporeSigma
  • Degradation testing of Mg alloys in Dulbecco's modified eagle medium: Influence of medium sterilization.
Degradation testing of Mg alloys in Dulbecco's modified eagle medium: Influence of medium sterilization.
Materials science & engineering. C, Materials for biological applications (2016-03-10)
Iñigo Marco, Frank Feyerabend, Regine Willumeit-Römer, Omer Van der Biest
ABSTRACT
This work studies the in vitro degradation of Mg alloys for bioabsorbable implant applications under near physiological conditions. For this purpose, the degradation behaviour of Mg alloys in Dulbecco's modified eagle medium (DMEM) which is a commonly used cell culture medium is analysed. Unfortunately, DMEM can be contaminated by microorganisms, acidifying the medium and accelerating the Mg degradation process by dissolution of protective degradation layers, such as (Mgx,Cay)(PO4)z. In this paper the influence of sterilization by applying UV-C radiation and antibiotics (penicillin/streptomycin) is analysed with two implant material candidates: Mg-Gd and Mg-Ag alloys; and pure magnesium as well as Mg-4Y-3RE as a reference.
Sigma-Aldrich
Dulbecco′s Modified Eagle′s Medium/Nutrient Mixture F-12 Ham, With L-glutamine, 15 mM HEPES, and sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture