MilliporeSigma
  • A quick and effective methodology for analyzing dinotefuran and its highly polar metabolites in plum using liquid chromatography-tandem mass spectrometry.
A quick and effective methodology for analyzing dinotefuran and its highly polar metabolites in plum using liquid chromatography-tandem mass spectrometry.
Food chemistry (2017-09-07)
Md Musfiqur Rahman, A M Abd El-Aty, Md Humayun Kabir, Hyung Suk Chung, Han Sol Lee, Fazil Hacımüftüoğlu, Ji Hoon Jeong, Byung-Joon Chang, Ho-Chul Shin, Jae-Han Shim
ABSTRACT
A simple and effective method was developed for analyzing dinotefuran and its three metabolites (MNG, UF, and DN) in plum using liquid chromatography-tandem mass spectrometry (LC-MS/MS). Due to the polarity and high water miscibility, dinotefuran and some of its metabolites (especially DN) have some limitations to be extracted with acetonitrile and salt following the "QuEChERS" sample preparation methodology. Alternatively, the samples were extracted with methanol, and purified with dispersive-solid phase extraction procedure (d-SPE) using primary secondary amine (PSA) and C18 sorbents after filtration, and mass up. Due to the suppression effect originated from plum matrix, matrix-matched calibration curves, which provided good linearity with coefficient of determination (R2)≥0.998, were used for quantification of all analytes. Blank plum samples fortified with 2 spiking levels (10×LOQ and 50×LOQ) yielded satisfactory recoveries for all tested analytes in the range of 83.01 to 110.18% with relative standard deviation (RSD)≤8.91. The method was successfully applied to field-incurred plum samples and dinotefuran and all metabolites were positively detected and quantified. In conclusion, we suggest that the method can be expanded to polar compounds having solvent and partitioning problems in any of the versions of QuEChERS.
Product Description
Supelco
Dinotefuran, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Pricing and availability is not currently available.