MilliporeSigma
  • Functionality of Redox-Active Cysteines Is Required for Restriction of Retroviral Replication by SAMHD1.
Functionality of Redox-Active Cysteines Is Required for Restriction of Retroviral Replication by SAMHD1.
Cell reports (2018-07-26)
Zhonghua Wang, Akash Bhattacharya, Tommy White, Cindy Buffone, Aine McCabe, Laura A Nguyen, Caitlin N Shepard, Sammy Pardo, Baek Kim, Susan T Weintraub, Borries Demeler, Felipe Diaz-Griffero, Dmitri N Ivanov
ABSTRACT
SAMHD1 is a dNTP triphosphohydrolase (dNTPase) that impairs retroviral replication in a subset of non-cycling immune cells. Here we show that SAMHD1 is a redox-sensitive enzyme and identify three redox-active cysteines within the protein: C341, C350, and C522. The three cysteines reside near one another and the allosteric nucleotide binding site. Mutations C341S and C522S abolish the ability of SAMHD1 to restrict HIV replication, whereas the C350S mutant remains restriction competent. The C522S mutation makes the protein resistant to inhibition by hydrogen peroxide but has no effect on the tetramerization-dependent dNTPase activity of SAMHD1 in vitro or on the ability of SAMHD1 to deplete cellular dNTPs. Our results reveal that enzymatic activation of SAMHD1 via nucleotide-dependent tetramerization is not sufficient for the establishment of the antiviral state and that retroviral restriction depends on the ability of the protein to undergo redox transformations.
Sigma-Aldrich
Iodoacetamide-13C2, 2-d2, 99 atom % 13C, 98 atom % D
Pricing and availability is not currently available.