MilliporeSigma
  • Synthesis and evaluation of an N-acylated photoactivatable analogue of glutathione as probe for glutathione-utilizing enzymes.
Synthesis and evaluation of an N-acylated photoactivatable analogue of glutathione as probe for glutathione-utilizing enzymes.
Bioorganic & medicinal chemistry letters (2006-01-03)
Dan Bernardi, Eric Battaglia, Gilbert Kirsch
ABSTRACT
The first synthesis of an N-acylated photoactivatable analogue of reduced glutathione is described. N-(4-Benzoylbenzoyl)glutathione (8) was found to be an inhibitor and a photoaffinity probe of purified rat liver glutathione S-transferases.