MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Phase I study of TAS-102 treatment in Japanese patients with advanced solid tumours.
Phase I study of TAS-102 treatment in Japanese patients with advanced solid tumours.
British journal of cancer (2012-06-28)
T Doi, A Ohtsu, T Yoshino, N Boku, Y Onozawa, A Fukutomi, S Hironaka, W Koizumi, T Sasaki
ABSTRACT
TAS-102 consists of α, α, α-trifluorothymidine (TFT) and an inhibitor of thymidine phosphorylase (TPI). We conducted a dose-escalation phase I study in Japanese patients with advanced solid tumours. TAS-102 was administered twice daily on days 1-5 and days 8-12 in a 28-day cycle to patients with solid tumours refractory to standard chemotherapy, to determine its maximum tolerated dose (MTD), dose-limiting toxicities (DLTs), and pharmacokinetics (PKs). MTD was evaluated in cycle 1. Safety and PKs were evaluated in 21 patients treated with TAS-102 at 30, 40, 50, 60, or 70 mg m(-2) per day. DLTs, such as grade 4 leucopenia, grade 4 neutropenia, and grade 4 thrombocytopenia, were observed in two patients at doses of 30 and 70 mg m(-2). α, α, α-trifluorothymidine and TPI exposures increased dose dependently, and the percentage of decrease in neutrophil count and TFT exposure were significantly correlated. The disease control rate was 50.0% with a median progression-free survival of 2.4 months in 18 colorectal cancer patients. The dose of TAS-102 was not increased above 70 mg m(-2) per day because of the increased tendency for grade 3 and 4 neutropenia, and 70 mg m(-2) per day was the recommended dose for phase II studies. TAS-102 at 70 mg m(-2) per day was tolerated in Japanese patients with advanced solid tumours. Phase II studies are ongoing in patients with colorectal cancer.
Sigma-Aldrich
Thymidine Phosphorylase, recombinant from Escherichia coli, recombinant, expressed in E. coli, buffered aqueous solution, ≥500 units/mL
Sigma-Aldrich
Thymidine Phosphorylase, recombinant from Escherichia coli, recombinant, expressed in E. coli, buffered aqueous solution, ≥900 units/mL, 0.2 μm filtered