MilliporeSigma
Single Molecule RNA FISH in Arabidopsis Root Cells.
Bio-protocol (2017-04-20)
Susan Duncan, Tjelvar S G Olsson, Matthew Hartley, Caroline Dean, Stefanie Rosa
ABSTRACT
Methods that allow the study of gene expression regulation are continually advancing. Here, we present an in situ hybridization protocol capable of detecting individual mRNA molecules in plant root cells, thus permitting the accurate quantification and localization of mRNA within fixed samples ( Duncan et al., 2016 ; Rosa et al., 2016 ). This single molecule RNA fluorescence in situ hybridization (smFISH) uses multiple single-labelled oligonucleotide probes to bind target RNAs and generate diffraction-limited signals that can be detected using a wide-field fluorescence microscope. We adapted a recent version of this method that uses 48 fluorescently labeled DNA oligonucleotides (20 mers) to hybridize to different portions of each transcript ( Raj et al., 2008 ). This approach is simple to implement and has the advantage that it can be readily applied to any genetic background.
Sigma-Aldrich
Catalase from bovine liver, aqueous solution, ≥30,000 units/mg protein