MilliporeSigma
  • Detection of 4a,5-dihydropravastatin as Impurity in the Cholesterol Lowering Drug Pravastatin.
Detection of 4a,5-dihydropravastatin as Impurity in the Cholesterol Lowering Drug Pravastatin.
Molecules (Basel, Switzerland) (2021-08-08)
Wibo B van Scheppingen, Peter P Lankhorst, Marcus Hans, Marco A van den Berg
ABSTRACT
Dihydro analogues are known byproducts of the fermentative production of statins and cannot be detected with existing pharmacopoeia analysis methods. We detected dihydropravastatin in most commercial formulations of pravastatin with LC-MS, in some cases in levels requiring identification. In fermentation broth samples of the single step production of pravastatin, we detected and identified for the first time 4a,5-dihydropravastatin, and confirmed that after several recrystallization steps this impurity can be fully removed from the pravastatin powder.
Sigma-Aldrich
Pravastatin sodium salt hydrate, ≥98% (HPLC), powder