MilliporeSigma
  • N-alpha-acetylation of Huntingtin protein increases its propensity to aggregate.
N-alpha-acetylation of Huntingtin protein increases its propensity to aggregate.
The Journal of biological chemistry (2021-11-05)
Leah Gottlieb, Lin Guo, James Shorter, Ronen Marmorstein
ABSTRACT
Huntington's disease (HD) is a neurodegenerative disorder caused by a poly-CAG expansion in the first exon of the HTT gene, resulting in an extended poly-glutamine tract in the N-terminal domain of the Huntingtin (Htt) protein product. Proteolytic fragments of the poly-glutamine-containing N-terminal domain form intranuclear aggregates that are correlated with HD. Post-translational modification of Htt has been shown to alter its function and aggregation properties. However, the effect of N-terminal Htt acetylation has not yet been considered. Here, we developed a bacterial system to produce unmodified or N-terminally acetylated and aggregation-inducible Htt protein. We used this system together with biochemical, biophysical, and imaging studies to confirm that the Htt N-terminus is an in vitro substrate for the NatA N-terminal acetyltransferase and show that N-terminal acetylation promotes aggregation. These studies represent the first link between N-terminal acetylation and the promotion of a neurodegenerative disease and implicates NatA-mediated Htt acetylation as a new potential therapeutic target in HD.
Sigma-Aldrich
Anti-polyQ specific Antibody, clone MW1, clone MW1, from mouse