MilliporeSigma
  • Protocol for reading and imaging live-cell PKA activity using ExRai-AKAR2.
Protocol for reading and imaging live-cell PKA activity using ExRai-AKAR2.
STAR protocols (2022-01-18)
Jin-Fan Zhang, Sohum Mehta, Jin Zhang
ABSTRACT
Fluorescent protein (FP)-based kinase activity biosensors are powerful tools for probing the spatiotemporal dynamics of signaling pathways in living cells. Yet, the limited sensitivity of most kinase biosensors restricts their reliable application in high-throughput detection modalities. Here, we report a protocol for using an ultrasensitive excitation-ratiometric PKA activity reporter, ExRai-AKAR2, to detect live-cell PKA activity via fluorescence microplate reading and epifluorescence microscopy. The high sensitivity of ExRai-AKAR2 is well suited to these high-throughput applications. For complete details on the use and execution of this protocol, please refer to Mehta et al. (2018) andZhang et al., 2021a) .
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
3-Isobutyl-1-methylxanthine, BioUltra, ≥99%