MilliporeSigma
  • An IL6-Adenosine Positive Feedback Loop between CD73+ γδTregs and CAFs Promotes Tumor Progression in Human Breast Cancer.
An IL6-Adenosine Positive Feedback Loop between CD73+ γδTregs and CAFs Promotes Tumor Progression in Human Breast Cancer.
Cancer immunology research (2020-08-28)
Guoming Hu, Pu Cheng, Jun Pan, Shimin Wang, Qiannan Ding, Zhou Jiang, Lu Cheng, Xuan Shao, Liming Huang, Jian Huang
ABSTRACT
The tumor microenvironment induces immunosuppression via recruiting and expanding suppressive immune cells such as regulatory T cells (Treg) to promote cancer progression. In this study, we documented that tumor-infiltrating CD73+ γδTregs were the predominant Tregs in human breast cancer and exerted more potent immunosuppressive activity than CD4+ or CD8+ Tregs. We further demonstrated that cancer-associated fibroblast (CAF)-derived IL6, rather than TGFβ1, induced CD73+ γδTreg differentiation from paired normal breast tissues via the IL6/STAT3 pathway to produce more adenosine and become potent immunosuppressive T cells. CD73+ γδTregs could in turn promote IL6 secretion by CAFs through adenosine/A2BR/p38MAPK signaling, thereby forming an IL6-adenosine positive feedback loop. CD73+ γδTreg infiltration also impaired the tumoricidal functions of CD8+ T cells and significantly correlated with worse prognosis of patients. The data indicate that the IL6-adenosine loop between CD73+ γδTregs and CAFs is important to promote immunosuppression and tumor progression in human breast cancer, which may be critical for tumor immunotherapy.
Sigma-Aldrich
STAT3 Inhibitor XI, STX-0119, The STAT3 Inhibitor XI, STX-0119 controls the biological activity of STAT3. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.
Pricing and availability is not currently available.