MilliporeSigma
  • Dubious effects of methadone as an "anticancer" drug on ovarian cancer cell-lines and patient-derived tumor-spheroids.
Dubious effects of methadone as an "anticancer" drug on ovarian cancer cell-lines and patient-derived tumor-spheroids.
Gynecologic oncology (2022-01-17)
Heidelinde Fiegl, Judith Hagenbuchner, Christiana Kyvelidou, Beata Seeber, Sieghart Sopper, Irina Tsibulak, Verena Wieser, Elisabeth Reiser, Julia Roessler, Kaisa Huhtinen, Olli Carpén, Walther Parson, Susanne Sprung, Christian Marth, Michael J Ausserlechner, Alain G Zeimet
ABSTRACT
The opioid agonist D,L-methadone exerts analgesic effects via the mu opioid receptor, encoded by OPRM1 and therefore plays a role in chronic pain management. In preclinical tumor-models D,L-methadone shows apoptotic and chemo-sensitizing effects and was therefore hyped as an off-label "anticancer" drug without substantiation from clinical trials. Its effects in ovarian cancer (OC) are completely unexplored. We analyzed OPRM1-mRNA expression in six cisplatin-sensitive, two cisplatin-resistant OC cell-lines, 170 OC tissue samples and 12 non-neoplastic control tissues. Pro-angiogenetic, cytotoxic and apoptotic effects of D,L-methadone were evaluated in OC cell-lines and four patient-derived tumor-spheroid models. OPRM1 was transcriptionally expressed in 69% of OC-tissues and in three of eight OC cell-lines. D,L-methadone exposure significantly reduced cell-viability in five OC cell-lines irrespective of OPRM1 expression. D,L-methadone, applied alone or combined with cisplatin, showed no significant effects on apoptosis or VEGF secretion in cell-lines. Notably, in two of the four spheroid models, treatment with D,L-methadone significantly enhanced cell growth (by up to 121%), especially after long-term exposure. This is consistent with the observed attenuation of the inhibitory effects of cisplatin in three spheroid models when adding D,L-methadone. The effect of methadone treatment on VEGF secretion in tumor-spheroids was inconclusive. Our study demonstrates that certain OC samples express OPRM1, which, however, is not a prerequisite for D,L-methadone function. As such, D,L-methadone may exert also detrimental effects by stimulating the growth of certain OC-cells and abrogating cisplatin's therapeutic effect.
Sigma-Aldrich
Anti-OPRM1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Pricing and availability is not currently available.