MilliporeSigma
Interleukin-21: a multifunctional regulator of immunity to infections.
Microbes and infection (2010-09-08)
John S Yi, Maureen A Cox, Allan J Zajac
ABSTRACT
Interleukin-21 (IL-21) is a cytokine that has broad effects on both innate and adaptive immune responses. The roles of IL-21 in determining immunity to infections are currently being defined, and notably, it has been shown that IL-21 is most critical for sustaining T cell responses during chronic viral infections. This article discusses our current understanding of the immunobiology of IL-21, as well as its known and potential roles in influencing immunity to infections.