MilliporeSigma
Tryptophan-kynurenine ratio as a biomarker of bladder cancer.
BJU international (2020-08-15)
Sze Han Lee, Ratha Mahendran, Sin Mun Tham, Thomas Paulraj Thamboo, Billy Jianhao Chionh, Yi Xin Lim, Woon Chau Tsang, Qing Hui Wu, Jun Yang Chia, Melissa Hui Wen Tay, Benjamin Yen Seow Goh, Kelven Weijing Chen, Jeane Zepeda Mallari, Revathi Periaswami, Lata Raman, Shoa Nian Choo, Dorinda Yan Qin Kioh, Edmund Chiong, Kesavan Esuvaranathan, Eric Chun Yong Chan
ABSTRACT
To investigate plasma and urinary kynurenine (KYN)-tryptophan (TRP) ratios in bladder cancer, expression of indoleamine 2,3-dioxygenase 1 (IDO1) in relation to tryptophan 2,3-dioxygenase (TDO2) in bladder tumour, and the correlation of KYN-TRP ratio with bladder tumour burden. Metabotyping of the TRP-KYN metabolic axis was performed via a clinical case-control study. Expression of IDO1 and TDO2 was measured in human biopsied tissues. Correlational experiments between KYN-TRP ratio and bladder tumour were performed using a murine orthotopic prostate-specific antigen (PSA)-secreting MB49 bladder cancer model. We established for the first time that plasma TRP level was significantly decreased, while both plasma and urinary KYN-TRP ratios were significantly higher in bladder cancer patients, and expression level of IDO1 but not TDO2 was increased in human bladder tumour. We reported the positive correlation between IDO1 expression, KYN-TRP ratio, normalized PSA to creatinine, and bladder tumour burden in the murine model. Kynurenine-tryptophan ratio is a promising surveillance biomarker for bladder cancer, but would require further validation before clinical translation.
Sigma-Aldrich
Anti-IDO1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab3