MilliporeSigma
  • PRR11 induces filopodia formation and promotes cell motility via recruiting ARP2/3 complex in non-small cell lung cancer cells.
PRR11 induces filopodia formation and promotes cell motility via recruiting ARP2/3 complex in non-small cell lung cancer cells.
Genes & diseases (2022-01-11)
Zhili Wei, Ru Wang, Xun Yin, Lian Zhang, Yunlong Lei, Ying Zhang, Yi Li, Jiaqian Wu, Youquan Bu, Guoxiang Jin, Chundong Zhang
ABSTRACT
Filopodia, a finger-like structure and actin-rich plasma-membrane protrusion at the leading edge of the cell, has important roles in cell motility. However, the mechanisms of filopodia generation are not well-understood via the actin-related protein 2/3 (ARP2/3) complex in Non-Small Cell Lung Cancer (NSCLC) cells. We previously have demonstrated that PRR11 associates with the ARP2/3 complex to regulate cytoskeleton-nucleoskeleton assembly and chromatin remodeling. In this study, we further demonstrate that PRR11 involves in filopodia formation, focal adhesion turnover and cell motility through ARP2/3 complex. Cell phenotype assays revealed that the silencing of PRR11 increased cellular size and inhibited cell motility in NSCLC cells. Mechanistically, PRR11 recruited and co-localized with Arp2 at the membrane protrusion to promote filopodia formation but not lamellipodia formation. Notably, PRR11 mutant deletion of the proline-rich region 2 (amino acid residues 185-200) abrogated the effect of filopodia formation. In addition, PRR11-depletion inhibited filopodial actin filaments assembly and increased the level of active integrin β1 in the cell surface, whereas reduced the phosphorylation level of focal adhesion kinase (FAKY397) to repress focal adhesion turnover and cell motility in NSCLC cells. Taken together, our findings indicate that PRR11 has critical roles in controlling filopodia formation, focal adhesion turnover and cell motility by recruiting ARP2/3 complex, thus dysregualted expression of PRR11 potentially facilitates tumor metastasis in NSCLC cells.
Sigma-Aldrich
Anti-PRR11 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution