MilliporeSigma
  • Thermorheological Characterization of Healthier Reduced-Fat Cocoa Butter Formulated by Substitution with a Hydroxypropyl Methylcellulose (HPMC)-Based Oleogel.
Thermorheological Characterization of Healthier Reduced-Fat Cocoa Butter Formulated by Substitution with a Hydroxypropyl Methylcellulose (HPMC)-Based Oleogel.
Foods (Basel, Switzerland) (2021-05-01)
María Dolores Alvarez, Susana Cofrades, María Espert, Ana Salvador, Teresa Sanz
ABSTRACT
Cocoa butter (CB) is a main ingredient in pastry due to the unique functional properties of its fat, which is high in saturated fatty acids (SFAs). However, excessive consumption of SFAs is associated with the occurrence of several chronic diseases. This study researched the partial or total replacement of CB by an oleogel (OG) formulated with a healthier lipid profile, for mixed systems that would allow a partial substitution of CB in confectionery products. The "emulsion-templated approach" was used to develop a sunflower oil-HPMC-based OG. Different CB:OG ratios were formulated increasing the percentage replacement of CB by OG from 50 to 100%. Rheological and textural properties were determined and compared with a CB control at 20 and 10 °C. Oil-binding capacity was also analyzed. The systems showed a solid-like behavior, with higher elastic than viscous modulus, which increased with CB concentration. Compared with 20 °C, at 10 °C there was an increase in connectivity, viscoelasticity, and consistency of the systems, in response to a more complete CB crystallization. The replaced systems also presented a better lipid profile than CB. This evidence suggests that formulated CB:OG system at 50:50 ratio could become useful as a CB equivalent in chocolate products.
Product Description
Supelco
PUFA No.2 (Animal Source), analytical standard