MilliporeSigma
  • Production of unnaturally linked chimeric proteins using a combination of sortase-catalyzed transpeptidation and click chemistry.
Production of unnaturally linked chimeric proteins using a combination of sortase-catalyzed transpeptidation and click chemistry.
Nature protocols (2013-08-31)
Martin D Witte, Christopher S Theile, Tongfei Wu, Carla P Guimaraes, Annet E M Blom, Hidde L Ploegh
ABSTRACT
Chimeric proteins, including bispecific antibodies, are biological tools with therapeutic applications. Genetic fusion and ligation methods allow the creation of N-to-C and C-to-N fused recombinant proteins, but not unnaturally linked N-to-N and C-to-C fusion proteins. This protocol describes a simple procedure for the production of such chimeric proteins, starting from correctly folded proteins and readily available peptides. By equipping the N terminus or C terminus of the proteins of interest with a set of click handles using sortase A, followed by a strain-promoted click reaction, unnatural N-to-N and C-to-C linked (hetero) fusion proteins are established. Examples of proteins that have been conjugated via this method include interleukin-2, interferon-α, ubiquitin, antibodies and several single-domain antibodies. If the peptides, sortase A and the proteins of interest are in hand, the unnaturally N-to-N and C-to-C fused proteins can be obtained in 3-4 d.
Sigma-Aldrich
Trifluoroacetic acid, ReagentPlus®, 99%