MilliporeSigma
  • The Crumbs3-Pals1 complex participates in the establishment of polarity in mammalian epithelial cells.
The Crumbs3-Pals1 complex participates in the establishment of polarity in mammalian epithelial cells.
Journal of cell science (2003-05-29)
Michael H Roh, Shuling Fan, Chia-Jen Liu, Ben Margolis
ABSTRACT
In Drosophila, the Crumbs-Stardust-Discs-lost complex is required during the establishment of polarized epithelia. Embryos that lack a component of this complex or overexpress Crumbs exhibit defects in epithelial morphogenesis. We recently cloned a novel mammalian epithelial Crumbs isoform, Crumbs3 (CRB3). CRB3 exists in a complex at tight junctions (TJs) with Pals1 and PATJ, the mammalian homologues of Stardust and Discs lost, respectively. Here, we observe that overexpression of CRB3 leads to delayed TJ formation in MDCK epithelial cell monolayers and disruption of polarity in MDCK cysts cultured in collagen. Both phenomena require the last four residues of CRB3. Next, we expressed, in MDCK cells, a dominant-negative Myc-Lin-2-Pals1 chimeric protein, where the PDZ domain of Lin-2 was replaced with that of Pals1. TJ and apical polarity defects were also observed in these cells. Collectively, this suggests that the CRB-Pals1 interaction is important for formation of TJs and polarized epithelia. These results provide insight into the function of the mammalian Crumbs complex during TJ formation and epithelial polarization.
Sigma-Aldrich
Monoclonal Anti-Uvomorulin/E-Cadherin antibody produced in rat, clone DECMA-1, ascites fluid, buffered aqueous solution
Pricing and availability is not currently available.
Sigma-Aldrich
Monoclonal Anti-Ezrin antibody produced in mouse, clone 3C12, ascites fluid
Pricing and availability is not currently available.