MilliporeSigma
  • Kinases of the Focal Adhesion Complex Contribute to Cardiomyocyte Specification.
Kinases of the Focal Adhesion Complex Contribute to Cardiomyocyte Specification.
International journal of molecular sciences (2021-10-14)
Sacha Robert, Marcus Flowers, Brenda M Ogle
ABSTRACT
Differentiation of pluripotent stem cells to cardiomyocytes is influenced by culture conditions including the extracellular matrices or similar synthetic scaffolds on which they are grown. However, the molecular mechanisms that link the scaffold with differentiation outcomes are not fully known. Here, we determined by immunofluorescence staining and mass spectrometry approaches that extracellular matrix (ECM) engagement by mouse pluripotent stem cells activates critical components of canonical wingless/integrated (Wnt) signaling pathways via kinases of the focal adhesion to drive cardiomyogenesis. These kinases were found to be differentially activated depending on type of ECM engaged. These outcomes begin to explain how varied ECM composition of in vivo tissues with development and in vitro model systems gives rise to different mature cell types, having broad practical applicability for the design of engineered tissues.
Sigma-Aldrich
ILK inhibitor, Cpd 22, The ILK inhibitor, Cpd 22 controls the biological activity of ILK. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.
Sigma-Aldrich
Glasgow Minimum Essential Medium, With sodium bicarbonate, without L-glutamine, liquid, sterile-filtered, suitable for cell culture