MilliporeSigma
  • Co-Expression with Membrane CMTM6/4 on Tumor Epithelium Enhances the Prediction Value of PD-L1 on Anti-PD-1/L1 Therapeutic Efficacy in Gastric Adenocarcinoma.
Co-Expression with Membrane CMTM6/4 on Tumor Epithelium Enhances the Prediction Value of PD-L1 on Anti-PD-1/L1 Therapeutic Efficacy in Gastric Adenocarcinoma.
Cancers (2021-10-24)
Ziqi Wang, Zhi Peng, Qiyao Liu, Zixia Guo, Merey Menatola, Jing Su, Ting Li, Qing Ge, Pingzhang Wang, Lin Shen, Rong Jin
ABSTRACT
Anti-PD-1/L1 immunotherapy has been intensively used in heavily treated population with advanced gastric adenocarcinoma. However, the immunotherapeutic efficacy is low even in PD-L1 positive patients. We aimed to establish a new strategy based on the co-expression of CMTM6/4 and PD-L1 for patient stratification before immunotherapy. By analyzing the data obtained from TCGA and single-cell RNA sequencing at the mRNA level, and 6-color multiplex immunofluorescence staining of tumor tissues in tissue array and 48-case pre-immunotherapy patients at the protein level, we found that CMTM6/4 and PD-L1 co-expressed in both epithelial and mesenchymal regions of gastric adenocarcinoma. The tumor tissues had higher levels of CMTM6/4 expression than their adjacent ones. A positive correlation was found between the expression of CMTM6/4 and the expression of PD-L1 in tumor epithelium. Epithelial co-expression of CMTM6/4 and PD-L1 in gastric tumor region was associated with shorter overall survival but better short-term response to anti-PD-1/L1 immunotherapy. Thus, we developed a predictive model and three pathological patterns based on the membrane co-expression of CMTM6/4 and PD-L1 in tumor epithelial cells for pre-immunotherapy patient screening in gastric adenocarcinoma.
Sigma-Aldrich
Anti-CMTM6 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution