MilliporeSigma
  • Rationally designed Human Cytomegalovirus gB nanoparticle vaccine with improved immunogenicity.
Rationally designed Human Cytomegalovirus gB nanoparticle vaccine with improved immunogenicity.
PLoS pathogens (2020-12-29)
Michela Perotti, Jessica Marcandalli, Davide Demurtas, Federica Sallusto, Laurent Perez
ABSTRACT
Human cytomegalovirus (HCMV) is the primary viral cause of congenital birth defects and causes significant morbidity and mortality in immune-suppressed transplant recipients. Despite considerable efforts in vaccine development, HCMV infection still represents an unmet clinical need. In recent phase II trials, a MF59-adjuvanted gB vaccine showed only modest efficacy in preventing infection. These findings might be attributed to low level of antibodies (Abs) with a neutralizing activity induced by this vaccine. Here, we analyzed the immunogenicity of each gB antigenic domain (AD) and demonstrated that domain I of gB (AD5) is the main target of HCMV neutralizing antibodies. Furthermore, we designed, characterized and evaluated immunogenic responses to two different nanoparticles displaying a trimeric AD5 antigen. We showed that mice immunization with nanoparticles induces sera neutralization titers up to 100-fold higher compared to those obtained with the gB extracellular domain (gBECD). Collectively, these results illustrate with a medically relevant example the advantages of using a general approach combining antigen discovery, protein engineering and scaffold presentation for modern development of subunit vaccines against complex pathogens.
Sigma-Aldrich
Complement sera from rabbit, lyophilized powder
Pricing and availability is not currently available.