MilliporeSigma
  • Mechanisms Underlying Vascular Endothelial Growth Factor Receptor Inhibition-Induced Hypertension: The HYPAZ Trial.
Mechanisms Underlying Vascular Endothelial Growth Factor Receptor Inhibition-Induced Hypertension: The HYPAZ Trial.
Hypertension (Dallas, Tex. : 1979) (2021-03-30)
Kaisa M Mäki-Petäjä, Adam McGeoch, Lucy L Yang, Annette Hubsch, Carmel M McEniery, Paul A R Meyer, Fraz Mir, Parag Gajendragadkar, Nicola Ramenatte, Gayathri Anandappa, Sara Santos Franco, Simon J Bond, Carola-Bibiane Schönlieb, Yoeri Boink, Christoph Brune, Ian B Wilkinson, Duncan I Jodrell, Joseph Cheriyan
ABSTRACT
[Figure: see text].
Sigma-Aldrich
Acetylcholinesterase Activity Assay Kit, sufficient for 100 colorimetric tests
Pricing and availability is not currently available.