MilliporeSigma
  • Cell-fate transition and determination analysis of mouse male germ cells throughout development.
Cell-fate transition and determination analysis of mouse male germ cells throughout development.
Nature communications (2021-11-27)
Jiexiang Zhao, Ping Lu, Cong Wan, Yaping Huang, Manman Cui, Xinyan Yang, Yuqiong Hu, Yi Zheng, Ji Dong, Mei Wang, Shu Zhang, Zhaoting Liu, Shuhui Bian, Xiaoman Wang, Rui Wang, Shaofang Ren, Dazhuang Wang, Zhaokai Yao, Gang Chang, Fuchou Tang, Xiao-Yang Zhao
ABSTRACT
Mammalian male germ cell development is a stepwise cell-fate transition process; however, the full-term developmental profile of male germ cells remains undefined. Here, by interrogating the high-precision transcriptome atlas of 11,598 cells covering 28 critical time-points, we demonstrate that cell-fate transition from mitotic to post-mitotic primordial germ cells is accompanied by transcriptome-scale reconfiguration and a transitional cell state. Notch signaling pathway is essential for initiating mitotic arrest and the maintenance of male germ cells' identities. Ablation of HELQ induces developmental arrest and abnormal transcriptome reprogramming of male germ cells, indicating the importance of cell cycle regulation for proper cell-fate transition. Finally, systematic human-mouse comparison reveals potential regulators whose deficiency contributed to human male infertility via mitotic arrest regulation. Collectively, our study provides an accurate and comprehensive transcriptome atlas of the male germline cycle and allows for an in-depth understanding of the cell-fate transition and determination underlying male germ cell development.
Sigma-Aldrich
Anti-KLF4 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution