MilliporeSigma
  • A Novel Mechanism for Activation of Myosin Regulatory Light Chain by Protein Kinase C-Delta in Drosophila.
A Novel Mechanism for Activation of Myosin Regulatory Light Chain by Protein Kinase C-Delta in Drosophila.
Genetics (2020-08-06)
Pooneh Vaziri, Danielle Ryan, Christopher A Johnston, Richard M Cripps
ABSTRACT
Myosin is an essential motor protein, which in muscle is comprised of two molecules each of myosin heavy-chain (MHC), the essential or alkali myosin light-chain 1 (MLC1), and the regulatory myosin light-chain 2 (MLC2). It has been shown previously that MLC2 phosphorylation at two canonical serine residues is essential for proper flight muscle function in Drosophila; however, MLC2 is also phosphorylated at additional residues for which the mechanism and functional significance is not known. We found that a hypomorphic allele of Pkcδ causes a flightless phenotype; therefore, we hypothesized that PKCδ phosphorylates MLC2. We rescued flight disability by duplication of the wild-type Pkcδ gene. Moreover, MLC2 is hypophosphorylated in Pkcδ mutant flies, but it is phosphorylated in rescued animals. Myosin isolated from Pkcδ mutant flies shows a reduced actin-activated ATPase activity, and MLC2 in these myosin preparations can be phosphorylated directly by recombinant human PKCδ. The flightless phenotype is characterized by a shortened and disorganized sarcomere phenotype that becomes apparent following eclosion. We conclude that MLC2 is a direct target of phosphorylation by PKCδ, and that this modification is necessary for flight muscle maturation and function.
Sigma-Aldrich
Actin from rabbit muscle, ≥85% (SDS-PAGE), lyophilized powder
Pricing and availability is not currently available.