MilliporeSigma
  • Contribution of telomere G-quadruplex stabilization to the inhibition of telomerase-mediated telomere extension by chemical ligands.
Contribution of telomere G-quadruplex stabilization to the inhibition of telomerase-mediated telomere extension by chemical ligands.
Journal of the American Chemical Society (2011-08-19)
Chang-yue Chen, Quan Wang, Jia-quan Liu, Yu-hua Hao, Zheng Tan
ABSTRACT
Inhibition of telomerase activity through stabilizing telomere G-quadruplex with small chemical ligands is emerging as a novel strategy for cancer therapy. For the large number of ligands that have been reported to inhibit telomerase activity, it is difficult to validate the contribution of G-quadruplex stabilization to the overall inhibition. Using a modified telomere repeat amplification protocol (TRAP) method to differentiate the telomere G-quadruplex independent effect from dependent ones, we analyzed several ligands that have high affinity and/or selectivity to telomere G-quadruplex. Our results show that these ligands effectively inhibited telomerase activity in the absence of telomere G-quadruplex. The expected G-quadruplex-dependent inhibition was only obvious for the cationic ligands at low K(+) concentration, but it dramatically decreased at physiological concentration of K(+). These observations demonstrate that the ligands are much more than G-quadruplex stabilizers with a strong G-quadruplex-irrelevant off-target effect. They inhibit telomerase via multiple pathways in which stabilization of telomere G-quadruplex may only make a minor or neglectable contribution under physiologically relevant conditions depending on the stability of telomere G-quadruplex under ligand-free conditions.