MilliporeSigma
  • Extracellular DNA released by glycine-auxotrophic Staphylococcus epidermidis small colony variant facilitates catheter-related infections.
Extracellular DNA released by glycine-auxotrophic Staphylococcus epidermidis small colony variant facilitates catheter-related infections.
Communications biology (2021-07-24)
Junlan Liu, Zhen Shen, Jin Tang, Qian Huang, Ying Jian, Yao Liu, Yanan Wang, Xiaowei Ma, Qian Liu, Lei He, Min Li
ABSTRACT
Though a definitive link between small colony variants (SCVs) and implant-related staphylococcal infections has been well-established, the specific underlying mechanism remains an ill-explored field. The present study analyzes the role SCVs play in catheter infection by performing genomic and metabolic analyses, as well as analyzing biofilm formation and impacts of glycine on growth and peptidoglycan-linking rate, on a clinically typical Staphylococcus epidermidis case harboring stable SCV, normal counterpart (NC) and nonstable SCV. Our findings reveal that S. epidermidis stable SCV carries mutations involved in various metabolic processes. Metabolome analyses demonstrate that two biosynthetic pathways are apparently disturbed in SCV. One is glycine biosynthesis, which contributes to remarkable glycine shortage, and supplementation of glycine restores growth and peptidoglycan-linking rate of SCV. The other is overflow of pyruvic acid and acetyl-CoA, leading to excessive acetate. SCV demonstrates higher biofilm-forming ability due to rapid autolysis and subsequent eDNA release. Despite a remarkable decline in cell viability, SCV can facilitate in vitro biofilm formation and in vivo survival of NC when co-infected with its normal counterparts. This work illustrates an intriguing strategy utilized by a glycine-auxotrophic clinical S. epidermidis SCV isolate to facilitate biofilm-related infections, and casts a new light on the role of SCV in persistent infections.
Sigma-Aldrich
Lectin from Triticum vulgaris (wheat), peroxidase conjugate, lyophilized powder