MilliporeSigma
  • SGK1 signaling promotes glucose metabolism and survival in extracellular matrix detached cells.
SGK1 signaling promotes glucose metabolism and survival in extracellular matrix detached cells.
Cell reports (2021-03-18)
Joshua A Mason, Jordan A Cockfield, Daniel J Pape, Hannah Meissner, Michael T Sokolowski, Taylor C White, José C Valentín López, Juan Liu, Xiaojing Liu, Inmaculada Martínez-Reyes, Navdeep S Chandel, Jason W Locasale, Zachary T Schafer
ABSTRACT
Loss of integrin-mediated attachment to extracellular matrix (ECM) proteins can trigger a variety of cellular changes that affect cell viability. Foremost among these is the activation of anoikis, caspase-mediated cell death induced by ECM detachment. In addition, loss of ECM attachment causes profound alterations in cellular metabolism, which can lead to anoikis-independent cell death. Here, we describe a surprising role for serum and glucocorticoid kinase-1 (SGK1) in the promotion of energy production when cells are detached. Our data demonstrate that SGK1 activation is necessary and sufficient for ATP generation during ECM detachment and anchorage-independent growth. More specifically, SGK1 promotes a substantial elevation in glucose uptake because of elevated GLUT1 transcription. In addition, carbon flux into the pentose phosphate pathway (PPP) is necessary to accommodate elevated glucose uptake and PPP-mediated glyceraldehyde-3-phosphate (G3P) is necessary for ATP production. Thus, our data show SGK1 as master regulator of glucose metabolism and cell survival during ECM-detached conditions.
Sigma-Aldrich
EMD638683, ≥98% (HPLC)
Sigma-Aldrich
Anti-SGK1 Antibody, Upstate®, from rabbit