MilliporeSigma
  • Linker histone H1FOO regulates the chromatin structure in mouse zygotes.
Linker histone H1FOO regulates the chromatin structure in mouse zygotes.
FEBS letters (2018-07-03)
Satoshi Funaya, Masatoshi Ooga, Masataka G Suzuki, Fugaku Aoki
ABSTRACT
The chromatin structure in one-cell-stage mouse embryos is extremely loose and becomes tighter at the two-cell stage. As linker histones are involved in higher-order chromatin structure, we examined the involvement of the linker histone variant H1foo in the change of chromatin looseness between the one- and two-cell stages. H1foo knockdown causes the chromatin structure to be tighter in the pronucleus and increases deposition of the histone H3 variant H3.1/3.2 in the peripheral region of the pronucleus in one-cell-stage embryos. The decrease in chromatin looseness at the two-cell stage is less after overexpressing H1foo. These results suggest that H1foo is involved in the change in chromatin structure via nuclear deposition of H3 variants between the one- and two-cell stages.
Sigma-Aldrich
Anti-Src Antibody, CT, clone NL19 | 04-772, clone NL19, Upstate®, from rabbit