MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Isolation and structural elucidation of proline-containing cyclopentapeptides from an endolichenic Xylaria sp.
Isolation and structural elucidation of proline-containing cyclopentapeptides from an endolichenic Xylaria sp.
Journal of natural products (2011-03-25)
Wen Wu, Huanqin Dai, Li Bao, Biao Ren, Jingcai Lu, Yuanming Luo, Liangdong Guo, Lixin Zhang, Hongwei Liu
ABSTRACT
Two new cyclic pentapeptides (1 and 2) and the known blazein (3), ganodesterone (4), ergosterin (5), cerevisterol (6), 24-methylcholesta-4,6,8(14),22-tetraen-3-one (7), 5,8-epidioxyergosta-6,22-dien-3-ol (8), 16-α-d-mannopyranosyloxyisopimar-7-en-19-oic acid (9), and 16-hydroxy isopimar-7-en-19-oic acid (10) have been isolated from the crude extract of an endolichenic Xylaria sp. The structures of 1 and 2 were elucidated primarily by NMR and MS methods. The absolute configurations of 1 and 2 were assigned using Marfey's method on their acid hydrolysate. Compounds 1-10 were evaluated for activity against fungi and for synergistic antifungal activity. Compound 1 showed synergistic antifungal activity against Candida albicans SC5314 with 0.004 μg/mL ketoconazole.
Sigma-Aldrich
Ergosterol, ≥75%