MilliporeSigma
  • Graphitic phosphorus coordinated single Fe atoms for hydrogenative transformations.
Graphitic phosphorus coordinated single Fe atoms for hydrogenative transformations.
Nature communications (2020-08-15)
Xiangdong Long, Zelong Li, Guang Gao, Peng Sun, Jia Wang, Bingsen Zhang, Jun Zhong, Zheng Jiang, Fuwei Li
ABSTRACT
Single-atom metal-nitrogen-carbon (M-N-C) catalysts have sparked intensive interests, however, the development of an atomically dispersed metal-phosphorus-carbon (M-P-C) catalyst has not been achieved, although molecular metal-phosphine complexes have found tremendous applications in homogeneous catalysis. Herein, we successfully construct graphitic phosphorus species coordinated single-atom Fe on P-doped carbon, which display outstanding catalytic performance and reaction generality in the heterogeneous hydrogenation of N-heterocycles, functionalized nitroarenes, and reductive amination reactions, while the corresponding atomically dispersed Fe atoms embedded on N-doped carbon are almost inactive under the same reaction conditions. Furthermore, we find that the catalytic activity of graphitic phosphorus coordinated single-atom Fe sharply decreased when Fe atoms were transformed to Fe clusters/nanoparticles by post-impregnation Fe species. This work can be of fundamental interest for the design of single-atom catalysts by utilizing P atoms as coordination sites as well as of practical use for the application of M-P-C catalysts in heterogeneous catalysis.
Sigma-Aldrich
Phytic acid solution, 50 % (w/w) in H2O
Pricing and availability is not currently available.